Tris(4-methylphenyl)phosphine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Tri-p-tolylphosphine, Tris(p-tolyl)phosphine
A151967-100G
A151967-10G
A151967-500G
A151967-1G
A151967-25G
A151967-5G
103804-326EA
5
USD
103804-326
103842-476
76783-398
76783-400
76783-402
77299-918
77318-064
77318-066
Tris(4-methylphenyl)phosphine 98%
Tris(4-methylphenyl)phosphine
Precaución: for Research Use Only
Formula:
C₂₁H₂₁P MW: 304.37 g/mol Boiling Pt: 399.8 °C (760 mm Hg) Melting Pt: 144…146 °C Flash Pt: 206.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008542 CAS Number: 1038-95-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...