Triphenylmethyl mercaptan 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Triphenylmethanethiol
A156446-1G
A156446-25G
A156446-5G
103804-884EA
6
USD
103804-884
103843-086
77318-866
77318-868
77318-870
Triphenylmethyl mercaptan 98%
Triphenylmethyl mercaptan
Precaución: for Research Use Only
Formula:
C₁₉H₁₆S MW: 276.40 g/mol Boiling Pt: 403.8 °C (760 mm Hg) Melting Pt: 104…106 °C Flash Pt: 166.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004854 CAS Number: 3695-77-0 UN: 3335 ADR: 9,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...