Triphenylmethylamine 98%

Supplier: Ambeed
Warning

Synonyms: Triphenylaminomethane, Tritylamine, Triphenylmethanamine

A206030-100G A206030-10G A206030-500G A206030-1000G A206030-25G
103809-424EA 167 USD
103809-424 103848-354 76785-502 76785-504 76785-506 77086-590 77302-978
Triphenylmethylamine 98%
Triphenylmethylamine

Precaución: for Research Use Only

Formula: C₁₉H₁₇N
MW: 259.35 g/mol
Boiling Pt: 369.3 °C (760 mm Hg)
Melting Pt: 102…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00008047
CAS Number: 5824-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR