Triphenylamine 97%

Supplier: Ambeed
Warning

A125855-100G A125855-10G A125855-500G A125855-5G A125855-25G
76782-004EA 50 USD
76782-004 76782-006 76782-008 77298-096 77312-884
Triphenylamine 97%
Triphenylamine

Precaución: for Research Use Only

Formula: C₁₈H₁₅N
Boiling Pt: 365 °C (760 mm Hg)
Melting Pt: 124…128 °C
Flash Pt: 365 °C
Storage Temperature: Ambient
MDL Number: MFCD00003020
CAS Number: 603-34-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR