Trimethylsulfoxonium bromide 98%

Supplier: Ambeed
Warning

A239311-1000G A239311-25G A239311-100G A239311-10G
103809-698EA 169 USD
103809-698 77090-216 77568-324 77593-534 77612-326
Trimethylsulfoxonium bromide 98%
Trimethylsulfoxonium bromide

Precaución: For research use only.

Formula: C₃H₉BrOS
MW: 173.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02093502
CAS Number: 25596-24-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR