3,4,5-Trimethoxybenzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: Trimethylgallic Acid

A341413-100G A341413-500G A341413-25G
103810-176EA 70 USD
103810-176 77277-984 77277-986 77306-532
3,4,5-Trimethoxybenzoic acid 98%
3,4,5-Trimethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₅
MW: 212.2 g/mol
Boiling Pt: 225…227 °C (760 mm Hg)
Melting Pt: 168…171 °C
Storage Temperature: Ambient
MDL Number: MFCD00002501
CAS Number: 118-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR