1-Phenyl-2-(trimethylsilyl)acetylene 95%

Supplier: Ambeed
Warning

Synonyms: Trimethyl(phenylethynyl)silane

A639556-5G A639556-100G A639556-10G A639556-1G
103840-336EA 25 USD
103840-336 76844-512 76980-736 76980-738 77308-042
1-Phenyl-2-(trimethylsilyl)acetylene 95%
1-Phenyl-2-(trimethylsilyl)acetylene

Precaución: For research use only

Formula: C₁₁H₁₄Si
MW: 174.32 g/mol
Boiling Pt: 217.4 °C (760 mm Hg)
Flash Pt: 73.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00054860
CAS Number: 2170-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR