Trimethyl methanetricarboxylate 98%

Supplier: Ambeed
Warning

A122659-250MG A122659-25G A122659-5G
103800-248EA 10 USD
103800-248 77026-528 77026-530 77312-378
Trimethyl methanetricarboxylate 98%
Trimethyl methanetricarboxylate

Precaución: for Research Use Only

Formula: C₈H₁₂O₆
MW: 204.18 g/mol
Boiling Pt: 242.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD02178862
CAS Number: 1186-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR