Trimesic acid trimethyl ester 95%

Supplier: Ambeed
Warning

Synonyms: Trimethyl 1,3,5-benzenetricarboxylate, Trimethyl benzene-1,3,5-tricarboxylate, 1,3,5-Benzenetricarboxylic acid trimethyl ester

A135907-100G A135907-10G A135907-500G A135907-25G A135907-5G
76719-178EA 72.15 USD
76719-178 76719-180 76719-182 77315-114 77315-116
Trimesic acid trimethyl ester 95%
Trimesic acid trimethyl ester

Precaución: for Research Use Only

Formula: C₁₂H₁₂O₆
MW: 252.22 g/mol
Boiling Pt: 332.8 °C (760 mm Hg)
Melting Pt: 144…147 °C
Flash Pt: 144.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00008434
CAS Number: 2672-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR