Octadecyltrimethoxysilane 90%
Supplier: Ambeed
|
Warning
|
Synonyms:
Trimethoxy(octadecyl)silane
A100921-10G
A100921-1G
A100921-25G
A100921-5G
103821-388EA
7
USD
103821-388
76717-506
76717-508
76717-510
77296-910
Octadecyltrimethoxysilane 90%
Octadecyltrimethoxysilane
Precaución: for Research Use Only
Formula:
C₂₁H₄₆O₃Si MW: 374.68 g/mol Melting Pt: 16…17 °C Flash Pt: 140 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043060 CAS Number: 3069-42-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...