1H,1H,2H,2H-Perfluorooctyltrimethoxysilane 95%

Supplier: Ambeed
Warning

A199618-25G A199618-5G A199618-100G A199618-10G A199618-1G
103847-742EA 6 USD
103847-742 77085-782 77085-784 77237-096 77237-098 77237-100
1H,1H,2H,2H-Perfluorooctyltrimethoxysilane 95%
1H,1H,2H,2H-Perfluorooctyltrimethoxysilane

Precaución: for Research Use Only

Formula: C₁₁H₁₃F₁₃O₃Si
Boiling Pt: 217.3 °C (760 mm Hg)
Melting Pt: <0 °C
Flash Pt: 85.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00153261
CAS Number: 85857-16-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR