Triisopropylsilyl trifluoromethanesulfonate 97% (by titrimetric analysis)

Supplier: Ambeed
Danger

Synonyms: Trifluoromethanesulphonic acid triisopropylsilyl ester, Tri(propan-2-yl)silyl trifluoromethanesulfonate

A274403-1G A274403-5G
103853-120EA 6 USD
103853-120 76787-252 77305-164
Triisopropylsilyl trifluoromethanesulfonate 97% (by titrimetric analysis)
Triisopropylsilyl trifluoromethanesulphonate

Precaución: for Research Use Only

Formula: C₁₀H₂₁F₃O₃SSi
MW: 306.42 g/mol
Boiling Pt: 252.9 °C (760 mm Hg)
Flash Pt: 100.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009913
CAS Number: 80522-42-5
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR