Triethyl methanetricarboxylate 97%

Supplier: Ambeed
Warning

A250776-100G A250776-10G A250776-500G A250776-25G A250776-5G
103851-790EA 9 USD
103851-790 76786-816 76786-818 76786-820 77091-154 77091-156
Triethyl methanetricarboxylate 97%
Triethyl methanetricarboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₆O₆
MW: 232.23 g/mol
Boiling Pt: 281.3 °C (760 mm Hg)
Melting Pt: 25…26 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00009150
CAS Number: 6279-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR