Triethyl-2-fluoro-2-phosphonoacetate (fluorinated horner reagent) 98%

Supplier: Ambeed
Warning

Synonyms: Diethyl [(ethoxycarbonyl)(fluoro)methyl]phosphonate, Triethyl 2-fluoro-2-phosphonoacetate

A414164-1G A414164-25G A414164-5G A414164-100G A414164-10G A414164-500G
103827-272EA 11 USD
103827-272 103838-316 76841-882 76841-884 76841-886 77551-238 77594-274 77616-806
Triethyl-2-fluoro-2-phosphonoacetate (fluorinated horner reagent) 98%
Triethyl 2-fluoro-2-phosphonoacetate (fluorinated Horner reagent)

Precaución: for Research Use Only

Formula: C₈H₁₆FO₅P
MW: 242.19 g/mol
Boiling Pt: 339.5 °C (760 mm Hg)
Flash Pt: 73.88 °C
Storage Temperature: Ambient
MDL Number: MFCD00134400
CAS Number: 2356-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR