Tridecane-1,13-diol 97%

Supplier: Ambeed
Warning

A410485-1G A410485-10G A410485-250MG A410485-5G
103827-212EA 7 USD
103827-212 103838-260 76841-780 77196-068 77196-070 77307-146
Tridecane-1,13-diol 97%
Tridecane-1,13-diol

Precaución: for Research Use Only

Formula: C₁₃H₂₈O₂
Boiling Pt: 343.9 °C (760 mm Hg)
Flash Pt: 154.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00482067
CAS Number: 13362-52-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR