Tricyclohexylphosphine 97%

Supplier: Ambeed
Warning

A583295-5G A583295-100G A583295-10G A583295-25G
103828-874EA 10 USD
103828-874 103840-024 76844-154 77103-812 77103-814 77307-874
Tricyclohexylphosphine 97%
Tricyclohexylphosphine

Precaución: for Research Use Only

Formula: C₁₈H₃₃P
Boiling Pt: 383.4 °C (760 mm Hg)
Melting Pt: 81…83 °C
Flash Pt: 8.888 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003853
CAS Number: 2622-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR