2,2,6,6-Tetramethyl-4-piperidone 97%

Supplier: Ambeed
Danger

Synonyms: Triacetonamine, 2,2,6,6-Tetramethylpiperidin-4-one

A112999-25G A112999-5G A112999-100G A112999-500G
103823-980EA 14 USD
103823-980 77025-856 77310-816 77592-204 77599-630
2,2,6,6-Tetramethyl-4-piperidone 97%
2,2,6,6-Tetramethyl-4-piperidone

Precaución: for Research Use Only

Formula: C₉H₁₇NO
MW: 155.24 g/mol
Boiling Pt: 102…105 °C (760 mm Hg)
Melting Pt: 34…38 °C
Flash Pt: 73 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00149393
CAS Number: 826-36-8
UN: 3259
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR