Tripyrrolidinophosphine oxide 98%
Supplier: Ambeed
|
Warning
|
A396022-25G
A396022-100G
A396022-10G
A396022-500G
A396022-1G
A396022-5G
103826-974EA
6
USD
103826-974
103838-012
76841-436
77115-692
77115-694
77115-696
77307-038
77307-040
Tripyrrolidinophosphine oxide 98%
Tripyrrolidinophosphine oxide
Precaución: for Research Use Only
Formula:
C₁₂H₂₄N₃OP Boiling Pt: 389.9 °C (760 mm Hg) Flash Pt: 112.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014095 CAS Number: 6415-07-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...