Tri(pyrrolidin-1-yl)phosphine 80%

Supplier: Ambeed
Warning

A141521-10G A141521-1G A141521-25G A141521-5G
103803-004EA 21 USD
103803-004 76984-218 77299-220 77299-222 77299-224
Tri(pyrrolidin-1-yl)phosphine 80%
Tri(pyrrolidin-1-yl)phosphine

Precaución: for Research Use Only

Formula: C₁₂H₂₄N₃P
Boiling Pt: 104 °C (0.1 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD01863555
CAS Number: 5666-12-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR