trans-1-Boc-4-fluoro-3-hydroxypiperidine 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
trans-tert-Butyl 4-fluoro-3-hydroxypiperidine-1-carboxylate
A266331-100MG
A266331-250MG
A266331-1G
103812-798EA
160
USD
103812-798
103852-700
77031-862
77031-864
77092-406
trans-1-Boc-4-fluoro-3-hydroxypiperidine 95%
trans-1-Boc-4-fluoro-3-hydroxypiperidine
Precaución: for Research Use Only
Formula:
C₁₀H₁₈FNO₃ MW: 219.26 g/mol Boiling Pt: 300.795 °C (760 mm Hg) Flash Pt: 135.717 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD18425630 CAS Number: 955028-82-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...