trans-N1,N1-Dimethylcyclohexane-1,4-diamine hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A189320-100MG
A189320-250MG
A189320-5G
A189320-1G
103816-178EA
514
USD
103816-178
77012-238
77012-240
77012-242
77084-540
trans-N1,N1-Dimethylcyclohexane-1,4-diamine hydrochloride 95%
trans-N1,N1-Dimethylcyclohexane-1,4-diamine hydrochloride
Precaución: for Research Use Only
Formula:
C₈H₁₉ClN₂ Storage Temperature: Ambient |
MDL Number:
MFCD24369756 CAS Number: 1388893-25-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...