Methyl trans-4-hydroxycyclohexanecarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: trans-4-Hydroxycyclohexanecarboxylic Acid Methyl Ester, trans-Methyl 4-hydroxycyclohexanecarboxylate

A168620-100G A168620-10G A168620-1G A168620-25G A168620-5G
103806-304EA 8 USD
103806-304 103844-558 76985-320 76985-322 77081-610 77081-612 77081-614
Methyl trans-4-hydroxycyclohexanecarboxylate 98%
Methyl trans-4-hydroxycyclohexanecarboxylate

Precaución: for Research Use Only

Formula: C₈H₁₄O₃
MW: 158.20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20441963
CAS Number: 6125-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR