trans-Methyl 4-aminocyclohexanecarboxylate 97%

Supplier: Ambeed
Warning

A112644-10G A112644-250MG A112644-1G
103798-060EA 32 USD
103798-060 103823-898 77233-644 77233-646 77310-712
trans-Methyl 4-aminocyclohexanecarboxylate 97%
trans-Methyl 4-aminocyclohexanecarboxylate

Precaución: for Research Use Only

Formula: C₈H₁₅NO₂
MW: 157.21 g/mol
Storage Temperature: Freezer
MDL Number: MFCD12913678
CAS Number: 62456-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR