trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid (trans-) 98%

Supplier: Ambeed
Warning

Synonyms: (2E)-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid, (E)-Ferulic acid, Trans-3-(4-hydroxy-3-methoxyphenyl)acrylic acid, trans-Ferulic acid

A200560-100G A200560-10G A200560-25G A200560-500G
103809-346EA 175 USD
103809-346 103847-844 76975-232 76975-234 77085-910 77302-690 77611-942
trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid (trans-) 98%
trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
Boiling Pt: 372.3 °C (760 mm Hg)
Melting Pt: 168…172 °C
Flash Pt: 150.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00004400
CAS Number: 537-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR