Diethyl-trans-1,2-cyclopropanedicarboxylate 98%

Supplier: Ambeed
Warning

A173653-100G A173653-10G A173653-250MG A173653-1G A173653-25G A173653-5G
103806-854EA 10 USD
103806-854 103845-154 76799-464 76799-466 76799-468 77082-416 77082-418 77082-420
Diethyl-trans-1,2-cyclopropanedicarboxylate 98%
Diethyl-trans-1,2-cyclopropanedicarboxylate

Precaución: for Research Use Only

Formula: C₉H₁₄O₄
Boiling Pt: 246.3 °C (760 mm Hg)
Flash Pt: 98.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00198002
CAS Number: 3999-55-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR