trans-1,4-Cyclohexanediol 97%

Supplier: Ambeed
Warning

Synonyms: trans-Cyclohexane-1,4-diol, trans-1,4-Dihydroxycyclohexane

A108117-100G A108117-10G A108117-250MG A108117-25G A108117-1G A108117-5G
103797-302EA 9 USD
103797-302 103823-078 76780-942 76780-944 76780-946 76780-948 77309-922 77309-924
trans-1,4-Cyclohexanediol 97%
trans-1,4-Cyclohexanediol

Precaución: for Research Use Only

Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 252.4 °C (760 mm Hg)
Melting Pt: 141…142 °C
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00063612
CAS Number: 6995-79-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR