trans-Cyclohexane-1,3-dicarboxylic acid 95%

Supplier: Ambeed
Warning

A112652-250MG A112652-25G A112652-1G A112652-5G
103798-064EA 36 USD
103798-064 103823-902 76821-940 76821-942 77310-714 77310-716
trans-Cyclohexane-1,3-dicarboxylic acid 95%
trans-Cyclohexane-1,3-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₈H₁₂O₄
MW: 172.178 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01311241
CAS Number: 2305-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR