trans-Cinnamic acid 98%

Supplier: Ambeed
Warning

Synonyms: Cinnamic acid (only trans), (2E)-2-Phenyl-2-propenoic acid, trans-Cinnamic acid

A303025-100G A303025-10G A303025-500G A303025-25G
103810-024EA 45 USD
103810-024 76787-786 76787-788 76787-790 77305-860
trans-Cinnamic acid 98%
trans-Cinnamic acid

Precaución: for Research Use Only

Formula: C₉H₈O₂
MW: 148.16 g/mol
Boiling Pt: 264.999 °C (760 mm Hg)
Melting Pt: 131…136 °C
Flash Pt: 189.53 °C
Storage Temperature: Ambient
MDL Number: MFCD00004369
CAS Number: 140-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR