Z-trans-4-Hydroxy-L-prolinol 97%

Supplier: Ambeed
Warning

A208375-1G A208375-250MG A208375-25G A208375-5G A208375-10G
103817-872EA 10 USD
103817-872 77086-874 77112-382 77112-384 77112-386 77615-750
Z-trans-4-Hydroxy-L-prolinol 97%
Z-trans-4-Hydroxy-L-prolinol

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO₄
Boiling Pt: 433.9 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD03427085
CAS Number: 95687-41-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR