trans-4-Methylcyclohexanecarboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: trans-4-Methyl-1-cyclohexanecarboxylic acid

A138769-100G A138769-25G A138769-5G
77190-308EA 35.02 USD
77190-308 77315-718 77315-720
trans-4-Methylcyclohexanecarboxylic acid 98%
trans-4-Methylcyclohexanecarboxylic acid

Precaución: for Research Use Only

Formula: C₈H₁₄O₂
MW: 142.2 g/mol
Boiling Pt: 247.5 °C (760 mm Hg)
Melting Pt: 109…111 °C
Flash Pt: 110.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00074943
CAS Number: 13064-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR