trans-4-Methoxycyclohexanecarboxylic acid 97%

Supplier: Ambeed
Warning

A115534-100MG A115534-250MG A115534-1G
103798-584EA 79 USD
103798-584 77009-142 77009-144 77311-260
trans-4-Methoxycyclohexanecarboxylic acid 97%
trans-4-Methoxycyclohexanecarboxylic acid

Precaución: for Research Use Only

Formula: C₈H₁₄O₃
MW: 158.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20488929
CAS Number: 73873-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR