trans-4-Hydroxycyclohexanecarboxylic acid 98%
Supplier: Ambeed
|
Warning
|
A132170-1G
A132170-25G
A132170-5G
A132170-10G
A132170-100G
103801-698EA
6
USD
103801-698
76782-406
77314-254
77314-256
77314-258
77592-518
77599-904
trans-4-Hydroxycyclohexanecarboxylic acid 98%
trans-4-Hydroxycyclohexanecarboxylic acid
Precaución: for Research Use Only
Formula:
C₇H₁₂O₃ Boiling Pt: 307.9 °C (760 mm Hg) Melting Pt: 145 °C Flash Pt: 154 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093477 CAS Number: 3685-26-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...