trans-4-Dimethylaminocrotonic acid hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: (E)-4-Dimethylaminocrotonic acid hydrochloride, (E)-4-(Dimethylamino)-2-butenoic acid hydrochloride, (E)-4-(Dimethylamino)but-2-enoic acid hydrochloride

A375909-1G A375909-25G A375909-5G A375909-100G A375909-10G A375909-250MG
103826-572EA 6 USD
103826-572 103837-596 76840-890 76840-892 76840-894 76876-586 76876-588 76876-590
trans-4-Dimethylaminocrotonic acid hydrochloride 97%
trans-4-Dimethylaminocrotonic acid hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₂ClNO₂
MW: 165.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03695466
CAS Number: 848133-35-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR