Monomethyl-trans-1,4-cyclohexanedicarboxylate 98%
Supplier: Ambeed
|
Warning
|
A360333-100G
A360333-10G
A360333-500G
A360333-25G
A360333-5G
A360333-1G
103826-250EA
5
USD
103826-250
103837-258
76788-558
76788-560
76788-562
76840-458
76840-460
77306-708
Monomethyl-trans-1,4-cyclohexanedicarboxylate 98%
Monomethyl-trans-1,4-cyclohexanedicarboxylate
Precaución: for Research Use Only
Formula:
C₉H₁₄O₄ Boiling Pt: 303.1 °C (760 mm Hg) Flash Pt: 118.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD01311247 CAS Number: 15177-67-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...