trans-4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: trans-4-(Boc-amino)cyclohexanecarboxylic Acid

A153649-100G A153649-10G A153649-1G A153649-25G A153649-5G A153649-500G
103804-544EA 6 USD
103804-544 103842-718 76689-454 76689-456 76689-458 77318-386 77318-388 77318-390 77877-018
trans-4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid 98%
trans-4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₄
Boiling Pt: 396.7 °C (760 mm Hg)
Flash Pt: 193.7 °C
Storage Temperature: Ambient
MDL Number: MFCD01862293
CAS Number: 53292-89-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR