trans-4-(4-Chlorophenyl)cyclohexane-1-carboxylic acid 98%

Supplier: Ambeed
Warning

A121296-100G A121296-10G A121296-25G A121296-500G A121296-5G
77284-814EA 52 USD
77284-814 77284-816 77284-818 77284-820 77312-178
trans-4-(4-Chlorophenyl)cyclohexane-1-carboxylic acid 98%
trans-4-(4-Chlorophenyl)cyclohexane-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₅ClO₂
Boiling Pt: 387.1 °C (760 mm Hg)
Melting Pt: 252…254 °C
Flash Pt: 187.9 °C
Storage Temperature: Ambient
MDL Number: MFCD06797626
CAS Number: 49708-81-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR