trans-3-Aminocyclopentanol hydrochloride 95%

Supplier: Ambeed
Warning

A196471-100MG A196471-250MG A196471-5G
103816-848EA 124 USD
103816-848 103847-460 77030-006 77030-008 77085-416
trans-3-Aminocyclopentanol hydrochloride 95%
trans-3-Aminocyclopentanol hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₂ClNO
MW: 137.6079 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22380009
CAS Number: 124555-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR