Racemic trans-2-Fluorocyclopropanecarboxylic acid 97%

Supplier: Ambeed
Danger

Synonyms: trans-2-Fluorocyclopropanecarboxylic acid

A278241-100MG A278241-250MG A278241-1G A278241-5G
103813-404EA 14 USD
103813-404 103853-344 76875-242 76875-244 77093-256 77093-258
Racemic trans-2-Fluorocyclopropanecarboxylic acid 97%
Racemic trans-2-Fluorocyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₄H₅FO₂
MW: 104.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11499062
CAS Number: 130340-04-4
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR