trans-2,6-Dimethylmorpholine 95%
Supplier: Ambeed
|
Warning
|
A285838-100MG
A285838-10G
A285838-250MG
A285838-1G
A285838-5G
103813-690EA
27
USD
103813-690
103834-826
76977-500
76977-502
76977-504
77305-482
77305-484
trans-2,6-Dimethylmorpholine 95%
trans-2,6-Dimethylmorpholine
Precaución: for Research Use Only
Formula:
C₆H₁₃NO MW: 115.174 g/mol Boiling Pt: 146.6 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD09953181 CAS Number: 6485-45-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...