trans-2,5-Dimethylpiperazine 95%

Supplier: Ambeed
Danger

A384302-5G A384302-10G A384302-25G
103837-798EA 6 USD
103837-798 76841-148 77003-668 77672-906
trans-2,5-Dimethylpiperazine 95%
trans-2,5-Dimethylpiperazine

Precaución: for Research Use Only

Formula: C₆H₁₄N₂
MW: 114.19 g/mol
Boiling Pt: 165.9 °C (760 mm Hg)
Melting Pt: 115…118 °C
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00070516
CAS Number: 2815-34-1
UN: 2926
ADR: 4.1,6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR