trans-2-(Benzylamino)cyclohexanol 97%

Supplier: Ambeed
Warning

Synonyms: Trans-2-(benzylamino)cyclohexanol

A109701-10G A109701-1G A109701-5G
103797-548EA 27 USD
103797-548 103823-354 77257-608 77310-158 77310-160
trans-2-(Benzylamino)cyclohexanol 97%
trans-2-(Benzylamino)cyclohexanol

Precaución: for Research Use Only

Formula: C₁₃H₁₉NO
MW: 205.30 g/mol
Boiling Pt: 339.2 °C (760 mm Hg)
Melting Pt: 69…72 °C
Flash Pt: 118.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00145421
CAS Number: 40571-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR