trans-2-(2-Methoxyphenyl)-5-oxotetrahydrofuran-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A288296-1G A288296-25G A288296-5G A288296-10G A288296-250MG
103813-788EA 7 USD
103813-788 103834-930 76837-486 76837-488 76837-490 77250-616 77250-618
trans-2-(2-Methoxyphenyl)-5-oxotetrahydrofuran-3-carboxylic acid 97%
trans-2-(2-Methoxyphenyl)-5-oxotetrahydrofuran-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₂O₅
MW: 236.22 g/mol
Boiling Pt: 479.1±45.0 °C (760 mm Hg)
Flash Pt: 190.6 °C
Storage Temperature: Ambient
MDL Number: MFCD09749700
CAS Number: 99226-02-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR