trans-1,4-Cyclohexanedimethanol 97%

Supplier: Ambeed
Warning

Synonyms: trans-1,4-Bis(hydroxymethyl)cyclohexane

A174797-1G A174797-25G A174797-5G A174797-100G A174797-10G A174797-500G
103806-974EA 5 USD
103806-974 103845-274 77082-578 77082-580 77082-582 77191-530 77191-532 77746-151
trans-1,4-Cyclohexanedimethanol 97%
trans-1,4-Cyclohexanedimethanol

Precaución: for Research Use Only

Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 286.2 °C (760 mm Hg)
Melting Pt: 67 °C
Flash Pt: 161.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00066360
CAS Number: 3236-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR