4-(trans-4-Butylcyclohexyl)cyclohexanone 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
trans-[4'-Butyl-1,1'-bicyclohexyl]-4-one
A198259-100G
A198259-10G
A198259-25G
A198259-500G
103817-004EA
8
USD
103817-004
103847-630
77085-646
77287-608
77287-610
77287-612
77287-614
77302-580
4-(trans-4-Butylcyclohexyl)cyclohexanone 95%
4-(trans-4-Butylcyclohexyl)cyclohexanone
Precaución: for Research Use Only
Formula:
C₁₆H₂₈O Boiling Pt: 327.8 °C (760 mm Hg) Flash Pt: 144.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD06797514 CAS Number: 92413-47-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...