3-Amino-1-propanesulfonic acid 98%
Supplier: Ambeed
|
Warning
|
A140624-100G
A140624-500G
A140624-25G
A140624-5G
77162-294EA
47
USD
77162-294
77162-296
77315-994
77315-996
3-Amino-1-propanesulfonic acid 98%
3-Amino-1-propanesulfonic acid
Precaución: for Research Use Only
Formula:
C₃H₉NO₃S Melting Pt: 270 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008225 CAS Number: 3687-18-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...