N-(Toluene-4-sulfonyl)glycine methyl ester 98%

Supplier: Ambeed
Warning

Synonyms: Methyl N-[(4-methylphenyl)sulphonyl]glycinate, Tos-Gly-Ome, N-Tosylglycine methyl ester, Methyl [(4-methylphenyl)sulphonamido]acetate

A185396-25G A185396-100G A185396-10G A185396-500G A185396-5G
103846-412EA 6 USD
103846-412 77084-052 77111-570 77111-572 77111-574 77301-966
N-(Toluene-4-sulfonyl)glycine methyl ester 98%
N-(Toluene-4-sulfonyl)glycine methyl ester

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₄S
MW: 243.28 g/mol
Boiling Pt: 240.5±23.0 °C (760 mm Hg)
Flash Pt: 101.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00558890
CAS Number: 2645-02-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR