1-Methyl-3-(3-methyl-4-(4-((trifluoromethyl)thio)phenoxy)phenyl)-1,3,5-triazinane-2,4,6-trione 98%

Supplier: Ambeed
Warning

Synonyms: Deltazuril, Toltrazuril

A477060-10G A477060-250MG A477060-1G A477060-5G A477060-100G
103828-092EA 19 USD
103828-092 103839-162 76818-586 76818-588 77307-522 77307-524 77673-232
1-Methyl-3-(3-methyl-4-(4-((trifluoromethyl)thio)phenoxy)phenyl)-1,3,5-triazinane-2,4,6-trione 98%
1-Methyl-3-(3-methyl-4-(4-((trifluoromethyl)thio)phenoxy)phenyl)-1,3,5-triazinane-2,4,6-trione

Precaución: for Research Use Only

Formula: C₁₈H₁₄F₃N₃O₄S
MW: 425.38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00867201
CAS Number: 69004-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR