N-(3-Chloro-2-methylphenyl)anthranilic acid 98%

Supplier: Ambeed
Danger

Synonyms: 2-[(3-Chloro-2-methylphenyl)amino]benzoic Acid, 2-(3-Chloro-o-toluidino)benzoic Acid, 2-((3-Chloro-2-methylphenyl)amino)benzoic acid, Tolfenamic Acid

A127860-100G A127860-10G A127860-1G A127860-25G A127860-5G
103801-028EA 13 USD
103801-028 76782-140 76782-142 77298-240 77298-242 77298-244
N-(3-Chloro-2-methylphenyl)anthranilic acid 98%
N-(3-Chloro-2-methylphenyl)anthranilic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₂ClNO₂
MW: 261.71 g/mol
Boiling Pt: 405.4 °C (760 mm Hg)
Melting Pt: 210…214 °C
Flash Pt: 199 °C
Storage Temperature: Ambient
MDL Number: MFCD00133865
CAS Number: 13710-19-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR