5-Chloro-6-((2-iminopyrrolidin-1-yl)methyl)pyrimidine-2,4(1H,3H)-dione hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: TPI Hydrochloride

A235932-100MG A235932-10MG A235932-250MG A235932-25G A235932-25MG A235932-50MG A235932-5MG A235932-1G A235932-5G
103811-046EA 74 USD
103811-046 103850-816 77193-202 77193-204 77193-206 77193-208 77193-210 77193-212 77193-214 77304-022 77304-024
5-Chloro-6-((2-iminopyrrolidin-1-yl)methyl)pyrimidine-2,4(1H,3H)-dione hydrochloride 98%
5-Chloro-6-((2-iminopyrrolidin-1-yl)methyl)pyrimidine-2,4(1H,3H)-dione hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₂Cl₂N₄O₂
MW: 279.12 g/mol
Melting Pt: 245 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01571384
CAS Number: 183204-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR