Tiotropium bromide ≥98%

Supplier: Ambeed
Danger

Synonyms: rel-(1R,2R,4S,5S,7s)-7-(2-Hydroxy-2,2-di(thiophen-2-yl)acetoxy)-9,9-dimethyl-3-oxa-9-azatricyclo[3.3.1.02,4]nonan-9-ium bromide

A166270-10MG A166270-1MG A166270-250MG A166270-50MG
103791-514EA 22 USD
103791-514 103792-256 103806-040 103844-300 77671-336 77671-338 77671-340 77671-342
Tiotropium bromide ≥98%
Tiotropium bromide

Precaución: Inert atmosphere, 2…8 °C.

Formula: C₁₉H₂₂BrNO₄S₂
MW: 472.41 g/mol
Melting Pt: 218…220 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00867027
CAS Number: 136310-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR